siRNA Details
| Gene Symbol: 4921539E11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163494.1 | NP_001156966.1 | 2 | 622 | ||
| NM_027612.1 | NP_081888.1 | 3 | 714 | |||
| GenBank mRNA | AK015018.1 | - | 3 | 709 | ||
| AK161421.1 | - | 3 | 714 | |||
| BC107404.1 | - | 2 | 563 | 40058776 | ||
| BC107405.1 | - | 3 | 655 | 40058778 | ||

4921539E11Rik
Genomic Map