siRNA Details
Gene Symbol: Mmrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163507.1 | NP_001156979.1 | 1, 2 | 827 | ||
NM_027613.1 | NP_081889.1 | 1, 2 | 827 | |||
GenBank mRNA | AK036724.1 | - | 1, 2 | 665 | ||
AK137556.1 | - | 1, 2 | 666 |
