siRNA Details
| Gene Symbol: 4922502B01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015021.1 | - | 244 | |||
| Gene Symbol: D030068K23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK083706.1 | - | 10 | 1519 | ||

4922502B01Rik
Genomic Map