siRNA Details
| Gene Symbol: Tex19.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027622.3 | NP_081898.1 | 3 | 1450 | ||
| GenBank mRNA | AK014985.1 | - | 3 | 1469 | ||
| BC141309.1 | - | 3 | 1040 | 9055921 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tex19.2| Gene Symbol: Tex19.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027622.3 | NP_081898.1 | 3 | 1450 | ||
| GenBank mRNA | AK014985.1 | - | 3 | 1469 | ||
| BC141309.1 | - | 3 | 1040 | 9055921 | ||