siRNA Details
| Gene Symbol: 4931431F19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027634.1 | NP_081910.1 | 1 | 658 | ||
| GenBank mRNA | AK016497.1 | - | 1 | 659 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4931431F19Rik| Gene Symbol: 4931431F19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027634.1 | NP_081910.1 | 1 | 658 | ||
| GenBank mRNA | AK016497.1 | - | 1 | 659 | ||