siRNA Details
Gene Symbol: 4931431F19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027634.1 | NP_081910.1 | 1 | 932 | ||
GenBank mRNA | AK016497.1 | - | 1 | 933 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4931431F19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027634.1 | NP_081910.1 | 1 | 932 | ||
GenBank mRNA | AK016497.1 | - | 1 | 933 |