siRNA Details
| Gene Symbol: Tmem30c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027651.1 | NP_081927.1 | 4 | 421 | ||
| GenBank mRNA | AK016597.1 | - | 4 | 435 | ||
| AK161475.1 | - | 3 | 2285 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem30c| Gene Symbol: Tmem30c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027651.1 | NP_081927.1 | 4 | 421 | ||
| GenBank mRNA | AK016597.1 | - | 4 | 435 | ||
| AK161475.1 | - | 3 | 2285 | |||