siRNA Details
| Gene Symbol: Zfp597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033159.2 | NP_001028331.2 | 1, 2 | 316 | ||
| GenBank mRNA | AK016727.1 | - | 1, 2 | 316 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp597| Gene Symbol: Zfp597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033159.2 | NP_001028331.2 | 1, 2 | 316 | ||
| GenBank mRNA | AK016727.1 | - | 1, 2 | 316 | ||