siRNA Details
| Gene Symbol: Sntg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290390.1 | NP_001277319.1 | 14 | 2682 | ||
| NM_001290392.1 | NP_001277321.1 | 11 | 2417 | |||
| NM_001290393.1 | NP_001277322.1 | 13 | 2631 | |||
| NM_027671.5 | NP_081947.2 | 14 | 2685 | |||
| GenBank mRNA | AF367759.1 | - | 13 | 1337 | ||
| AK038698.1 | - | 11 | 2418 | |||
| AK080034.1 | - | 14 | 2047 | |||
| AK083241.1 | - | 14 | 1670 | |||

Sntg1
Genomic Map