siRNA Details
| Gene Symbol: 4933407L21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016730.1 | - | 3 | 856 | ||
| AK037440.1 | - | 1 | 2581 | |||
| AW125058.1 | - | 1 | 303 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933407L21Rik| Gene Symbol: 4933407L21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016730.1 | - | 3 | 856 | ||
| AK037440.1 | - | 1 | 2581 | |||
| AW125058.1 | - | 1 | 303 | |||