siRNA Details
| Gene Symbol: Scara5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168318.1 | NP_001161790.1 | 4 | 753 | ||
| NM_028903.2 | NP_083179.2 | 4 | 753 | |||
| GenBank mRNA | AK016543.1 | - | 4 | 788 | ||
| AK016912.1 | - | 4 | 720 | |||
| AK033154.1 | - | 4 | 720 | |||
| AK133176.1 | - | 4 | 754 | |||
| BC016096.1 | - | 4 | 729 | 4506472 | ||
| BC023907.1 | - | 4 | 669 | 5321978 | ||
| BC032159.1 | - | 4 | 709 | 5354657 | ||
| DQ122126.1 | - | 3 | 403 | |||

Scara5
Genomic Map