siRNA Details
Gene Symbol: Golga7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141983.1 | NP_001135455.1 | 4 | 508 | ||
NM_027694.2 | NP_081970.1 | 2 | 499 | |||
GenBank mRNA | AK016850.1 | - | 4 | 513 | ||
AK139354.1 | - | 2 | 500 | |||
BC049116.1 | - | 2 | 455 | 6811056 |
