siRNA Details
| Gene Symbol: 4933416I08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027700.1 | NP_081976.1 | 860 | |||
| GenBank mRNA | AK016833.1 | - | 827 | |||
| AK163858.1 | - | 676 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933416I08Rik| Gene Symbol: 4933416I08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027700.1 | NP_081976.1 | 860 | |||
| GenBank mRNA | AK016833.1 | - | 827 | |||
| AK163858.1 | - | 676 | ||||