siRNA Details
Gene Symbol: Otud1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027715.1 | NP_081991.1 | 1 | 1171 | ||
GenBank mRNA | AK016972.1 | - | 1 | 478 | ||
AK206709.1 | - | 1 | 125 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Otud1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027715.1 | NP_081991.1 | 1 | 1171 | ||
GenBank mRNA | AK016972.1 | - | 1 | 478 | ||
AK206709.1 | - | 1 | 125 |