siRNA Details
| Gene Symbol: Gpr15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162955.1 | NP_001156427.1 | 1 | 24 | ||
| GenBank mRNA | BC107229.2 | - | 1 | 24 | 40052032 | |
| BC107281.2 | - | 1 | 22 | |||
| BC107282.2 | - | 1 | 22 | |||
| BC119501.1 | - | 1 | 24 | 40052770 | ||
| BC119502.1 | - | 1 | 24 | 40052772 | ||
| BC119518.1 | - | 1 | 24 | 40055788 | ||
More Information
Set Membership: |
|

Gpr15
Genomic Map