siRNA Details
Gene Symbol: Daw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027725.3 | NP_082001.2 | 3 | 226 | ||
GenBank mRNA | AK016201.1 | - | 3 | 200 | ||
AK016977.1 | - | 3 | 227 | |||
BC049704.1 | - | 3 | 226 | |||
BC144994.1 | - | 3 | 203 | |||
BC145995.1 | - | 3 | 226 | 40131298 | ||
BU612798.1 | - | 3 | 169 |
