siRNA Details
| Gene Symbol: Rsph14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163533.1 | NP_001157005.1 | 7 | 1060 | ||
| NM_001163534.1 | NP_001157006.1 | 7 | 1047 | |||
| NM_001163535.1 | NP_001157007.1 | 7 | 1044 | |||
| NM_027730.1 | NP_082006.1 | 7 | 1057 | |||
| GenBank mRNA | AK017008.1 | - | 7 | 1037 | ||
| BC147792.1 | - | 7 | 1016 | 9087509 | ||
| BC147801.1 | - | 7 | 1019 | 9087518 | ||

Rsph14
Genomic Map