siRNA Details
| Gene Symbol: Spata24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027733.5 | NP_082009.3 | 3 | 275 | ||
| NM_029485.2 | NP_083761.1 | 3 | 275 | |||
| GenBank mRNA | AK016341.1 | - | 3 | 281 | ||
| AK017218.1 | - | 3 | 251 | |||
| AY730532.1 | - | 3 | 275 | |||
| BC060950.1 | - | 3 | 228 | 6773733 | ||
| BM569619.1 | - | 4 | 417 | |||
| BU961109.1 | - | 3 | 278 | |||

Spata24
Genomic Map