siRNA Details
| Gene Symbol: Arhgap26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175164.4 | NP_780373.3 | 19, 20 | 2178 | ||
| GenBank mRNA | AK017029.1 | - | 2, 3 | 351 | ||
| AK129176.1 | - | 19, 20 | 2116 | |||
| AK152368.1 | - | 2, 3 | 195 | |||
| BC141394.1 | - | 19, 20 | 2047 | 9056007 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AW908925.1 | - | 465 | Chr. - - [-] on Build | ||

Arhgap26
Genomic Map