siRNA Details
| Gene Symbol: Arhgap26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175164.4 | NP_780373.3 | 22, 23 | 2677 | ||
| GenBank mRNA | AK129176.1 | - | 22, 23 | 2615 | ||
| AK152368.1 | - | 4, 5 | 418 | |||
| BC141394.1 | - | 21, 22 | 2270 | 9056007 | ||

Arhgap26
Genomic Map