siRNA Details
| Gene Symbol: Wdr31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290521.1 | NP_001277450.1 | 5, 6 | 630 | ||
| NM_001290522.1 | NP_001277451.1 | 5, 6 | 520 | |||
| NM_023597.2 | NP_076086.1 | 5, 6 | 523 | |||
| GenBank mRNA | AF274321.1 | - | 5, 6 | 500 | ||
| AK017259.1 | - | 5, 6 | 513 | |||
| AK141026.1 | - | 5, 6 | 628 | |||
| BC026500.1 | - | 4, 5 | 409 | |||
| BC037102.1 | - | 5, 6 | 513 | 5364307 | ||

Wdr31
Genomic Map