siRNA Details
Gene Symbol: Col24a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027770.2 | NP_082046.2 | 3 | 1976 | ||
GenBank mRNA | AY243578.1 | - | 1 | 1130 | ||
BC139134.1 | - | 3 | 1533 | 8862156 | ||
BC145544.1 | - | 3 | 1533 | |||
DQ157748.1 | - | 3 | 1976 |
