siRNA Details
| Gene Symbol: Col24a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027770.2 | NP_082046.2 | 3 | 2104 | ||
| GenBank mRNA | BC139134.1 | - | 3 | 1661 | 8862156 | |
| BC145544.1 | - | 3 | 1661 | |||
| DQ157748.1 | - | 3 | 2104 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Col24a1| Gene Symbol: Col24a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027770.2 | NP_082046.2 | 3 | 2104 | ||
| GenBank mRNA | BC139134.1 | - | 3 | 1661 | 8862156 | |
| BC145544.1 | - | 3 | 1661 | |||
| DQ157748.1 | - | 3 | 2104 | |||