siRNA Details
| Gene Symbol: Krtap7-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027771.1 | NP_082047.1 | 1 | 114 | ||
| GenBank mRNA | AK017387.1 | - | 1 | 115 | ||
| CB320982.1 | - | 1 | 97 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtap7-1| Gene Symbol: Krtap7-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027771.1 | NP_082047.1 | 1 | 114 | ||
| GenBank mRNA | AK017387.1 | - | 1 | 115 | ||
| CB320982.1 | - | 1 | 97 | |||