siRNA Details
| Gene Symbol: Pdss2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027772.2 | NP_082048.2 | 7, 8 | 1327 | ||
| GenBank mRNA | AB210840.1 | - | 7, 8 | 1051 | ||
| BC147693.1 | - | 7, 8 | 1104 | 9056666 | ||
| BC147694.1 | - | 7, 8 | 1104 | 9056667 | ||

Pdss2
Genomic Map