siRNA Details
| Gene Symbol: Pdss2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168289.1 | NP_001161761.1 | 2, 3 | 717 | ||
| NM_027772.2 | NP_082048.2 | 2, 3 | 717 | |||
| GenBank mRNA | AB210840.1 | - | 2, 3 | 441 | ||
| AK017329.1 | - | 2, 3 | 687 | |||
| AK140091.1 | - | 2, 3 | 604 | |||
| AK165732.1 | - | 2, 3 | 716 | |||
| BC147693.1 | - | 2, 3 | 494 | 9056666 | ||
| BC147694.1 | - | 2, 3 | 494 | 9056667 | ||
| BC147752.1 | - | 2, 3 | 467 | 9056701 | ||
| BC147753.1 | - | 2, 3 | 467 | 9056702 | ||
| BC172142.1 | - | 2, 3 | 420 | |||

Pdss2
Genomic Map