siRNA Details
| Gene Symbol: 5430416O09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017317.1 | - | 2 | 615 | ||
| AK030334.1 | - | 2 | 633 | |||
| BC064043.1 | - | 2 | 600 | |||
| BC147344.1 | - | 1 | 164 | 9056418 | ||
| BC147345.1 | - | 1 | 164 | 9056411 | ||

5430416O09Rik
Genomic Map