siRNA Details
| Gene Symbol: Fmnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172409.2 | NP_765997.2 | 17 | 2339 | ||
| GenBank mRNA | AF513716.1 | - | 6 | 736 | ||
| BC048004.1 | - | 2 | 227 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fmnl2| Gene Symbol: Fmnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172409.2 | NP_765997.2 | 17 | 2339 | ||
| GenBank mRNA | AF513716.1 | - | 6 | 736 | ||
| BC048004.1 | - | 2 | 227 | |||