siRNA Details
| Gene Symbol: Epg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195633.1 | NP_001182562.1 | 31 | 5434 | ||
| GenBank mRNA | AK122536.1 | - | 16 | 2418 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Epg5| Gene Symbol: Epg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195633.1 | NP_001182562.1 | 31 | 5434 | ||
| GenBank mRNA | AK122536.1 | - | 16 | 2418 | ||