siRNA Details
| Gene Symbol: 5530601H04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017459.1 | - | 409 | |||
| AK153718.1 | - | 284 | ||||
| AK164650.1 | - | 4, 5 | 473 | |||
| BC019699.1 | - | 287 | ||||
| BC049927.1 | - | 261 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5530601H04Rik| Gene Symbol: 5530601H04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017459.1 | - | 409 | |||
| AK153718.1 | - | 284 | ||||
| AK164650.1 | - | 4, 5 | 473 | |||
| BC019699.1 | - | 287 | ||||
| BC049927.1 | - | 261 | ||||