siRNA Details
| Gene Symbol: Mettl13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144877.1 | NP_659126.1 | 3 | 1120 | ||
| GenBank mRNA | AK048636.1 | - | 2 | 2685 | ||
| AK050492.1 | - | 3 | 1313 | |||
| AK084750.1 | - | 3 | 820 | |||
| AK146751.1 | - | 1318 | ||||
| AK162843.1 | - | 3 | 1305 | |||
| AK168959.1 | - | 3 | 1327 | |||
| AK169319.1 | - | 1319 | ||||
| AK173044.1 | - | 1273 | ||||
| BC014872.1 | - | 3 | 1120 | 3602469 | ||

Mettl13
Genomic Map