siRNA Details
| Gene Symbol: Fam217b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081289.1 | NP_001074758.1 | 4 | 847 | ||
| GenBank mRNA | AK155442.1 | - | 4 | 849 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam217b| Gene Symbol: Fam217b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081289.1 | NP_001074758.1 | 4 | 847 | ||
| GenBank mRNA | AK155442.1 | - | 4 | 849 | ||