siRNA Details
| Gene Symbol: 4930506M07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114312.1 | NP_001107784.1 | 4, 5 | 814 | ||
| NM_175172.4 | NP_780381.1 | 4, 5 | 814 | |||
| GenBank mRNA | AK082304.1 | - | 4, 5 | 815 | ||
| AK220405.1 | - | 4, 5 | 844 | |||
| BC030338.1 | - | 4, 5 | 386 | 2599101 | ||
| EF055486.1 | - | 4, 5 | 257 | |||

4930506M07Rik
Genomic Map