siRNA Details
| Gene Symbol: Rnf215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027859.2 | NP_082135.2 | 4 | 653 | ||
| GenBank mRNA | AK002414.1 | - | 4 | 611 | ||
| AK047194.1 | - | 602 | ||||
| AK154140.1 | - | 4 | 636 | |||
| AK183370.1 | - | 3 | 261 | |||
| AK206501.1 | - | 3 | 261 | |||
| BC103627.1 | - | 4 | 577 | 6486672 | ||
| BC115907.1 | - | 4 | 600 | 40105152 | ||
| BC115908.1 | - | 4 | 600 | 40105148 | ||

Rnf215
Genomic Map