siRNA Details
| Gene Symbol: 0610010F05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027860.2 | NP_082136.2 | 11 | 1464 | ||
| GenBank mRNA | AK029538.1 | - | 11 | 1385 | ||
| AK129454.1 | - | 11 | 1431 | |||
| BC055049.1 | - | 10 | 1337 | |||
| BC065122.1 | - | 12 | 1488 | |||
| BC072622.1 | - | 11 | 1454 | 6821718 | ||
| BC079876.1 | - | 11 | 1450 | 6848269 | ||

0610010F05Rik
Genomic Map