siRNA Details
| Gene Symbol: Tmem25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027865.2 | NP_082141.1 | 4 | 582 | ||
| GenBank mRNA | AK002841.1 | - | 4 | 583 | ||
| AK090375.1 | - | 3 | 988 | |||
| AK154206.1 | - | 3 | 1068 | |||
| AK165199.1 | - | 4 | 614 | |||
| AK165827.1 | - | 4 | 567 | |||
| BC060057.1 | - | 4 | 602 | 5696102 | ||
| BC060243.1 | - | 4 | 602 | 5696102 | ||

Tmem25
Genomic Map