siRNA Details
| Gene Symbol: Dram1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027878.2 | NP_082154.2 | 2 | 337 | ||
| GenBank mRNA | AK004552.1 | - | 2 | 337 | ||
| AK151184.1 | - | 2 | 263 | |||
| BC021433.1 | - | 2 | 303 | 5030012 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dram1| Gene Symbol: Dram1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027878.2 | NP_082154.2 | 2 | 337 | ||
| GenBank mRNA | AK004552.1 | - | 2 | 337 | ||
| AK151184.1 | - | 2 | 263 | |||
| BC021433.1 | - | 2 | 303 | 5030012 | ||