siRNA Details
| Gene Symbol: Cdc40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027879.2 | NP_082155.1 | 9, 10 | 1053 | ||
| GenBank mRNA | AK004569.1 | - | 9, 10 | 1020 | ||
| AK136292.1 | - | 1, 2 | 973 | |||
| BC147503.1 | - | 9, 10 | 1053 | 9056386 | ||
| BC158078.1 | - | 9, 10 | 1053 | 9088170 | ||

Cdc40
Genomic Map