siRNA Details
| Gene Symbol: Cic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF363690.1 | - | 2, 3 | 266 | ||
| AK147276.1 | - | 2, 3 | 899 | |||
| AK147535.1 | - | 3, 4 | 3017 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cic| Gene Symbol: Cic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF363690.1 | - | 2, 3 | 266 | ||
| AK147276.1 | - | 2, 3 | 899 | |||
| AK147535.1 | - | 3, 4 | 3017 | |||