siRNA Details
| Gene Symbol: Susd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027890.5 | NP_082166.3 | 3 | 390 | ||
| GenBank mRNA | AK004703.2 | - | 3 | 360 | ||
| AK028817.1 | - | 3 | 366 | |||
| AK037059.1 | - | 3 | 372 | |||
| AK037127.1 | - | 3 | 341 | |||
| AK038944.1 | - | 3 | 391 | |||
| AK042469.1 | - | 3 | 374 | |||
| AK128959.1 | - | 3 | 362 | |||
| AK167155.1 | - | 3 | 391 | |||
| BB632717.1 | - | 3 | 391 | |||
| BC026837.1 | - | 3 | 343 | 4222578 | ||

Susd2
Genomic Map