siRNA Details
Gene Symbol: Ulk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027895.1 | NP_082171.1 | 7 | 874 | ||
GenBank mRNA | AK004783.1 | - | 5 | 1456 | ||
AK032030.1 | - | 5 | 496 | |||
AK154642.1 | - | 7 | 787 | |||
AK186089.1 | - | 3 | 181 | |||
BC037093.1 | - | 7 | 895 | |||
BC039958.1 | - | 3 | 236 | |||
BC151153.1 | - | 7 | 800 | 9088047 |
