siRNA Details
| Gene Symbol: Cyp2d40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023623.2 | NP_076112.2 | 3 | 446 | ||
| GenBank mRNA | AK004984.1 | - | 3 | 509 | ||
| BC061152.1 | - | 3 | 446 | 30308952 | ||
| BC087906.1 | - | 3 | 507 | 30306972 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp2d40| Gene Symbol: Cyp2d40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023623.2 | NP_076112.2 | 3 | 446 | ||
| GenBank mRNA | AK004984.1 | - | 3 | 509 | ||
| BC061152.1 | - | 3 | 446 | 30308952 | ||
| BC087906.1 | - | 3 | 507 | 30306972 | ||