siRNA Details
| Gene Symbol: C2cd2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027909.2 | NP_082185.2 | 1, 2 | 758 | ||
| GenBank mRNA | AK004920.1 | - | 1, 2 | 763 | ||
| AK039220.1 | - | 1, 2 | 729 | |||
| AK122237.1 | - | 1, 2 | 704 | |||
| AK153804.1 | - | 1, 2 | 730 | |||
| AK189552.1 | - | 1, 2 | 70 | |||
| AK216579.1 | - | 1, 2 | 70 | |||
| BC056612.1 | - | 1, 2 | 329 | 4952570 | ||
| BC060156.1 | - | 1, 2 | 622 | 6810220 | ||

C2cd2l
Genomic Map