siRNA Details
| Gene Symbol: Tysnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001272090.1 | NP_001259019.1 | 1 | 300 | ||
| NM_001272091.1 | NP_001259020.1 | 1 | 300 | |||
| NM_001272092.1 | NP_001259021.1 | 1 | 300 | |||
| NM_027912.1 | NP_082188.1 | 1 | 300 | |||
| GenBank mRNA | AK005069.1 | - | 1 | 301 | ||
| AK154513.1 | - | 1 | 283 | |||
| BC020273.1 | - | 1 | 291 | |||
| BC119320.1 | - | 1 | 278 | 8734069 | ||

Tysnd1
Genomic Map