siRNA Details
| Gene Symbol: Bbs10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027914.1 | NP_082190.1 | 2 | 2063 | ||
| GenBank mRNA | AK004937.1 | - | 1 | 2369 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bbs10| Gene Symbol: Bbs10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027914.1 | NP_082190.1 | 2 | 2063 | ||
| GenBank mRNA | AK004937.1 | - | 1 | 2369 | ||