siRNA Details
| Gene Symbol: Pitpnc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145823.2 | NP_665822.1 | 5 | 1099 | ||
| GenBank mRNA | AB077281.1 | - | 5 | 1080 | ||
| AB077282.1 | - | 5 | 377 | |||
| AK003844.1 | - | 4 | 347 | |||
| AK021206.1 | - | 5 | 312 | |||
| AK153625.1 | - | 5 | 1100 | |||
| AK171340.1 | - | 5 | 1126 | |||
| AK171918.1 | - | 5 | 1096 | |||
| AK172137.1 | - | 5 | 1100 | |||
| BC082333.1 | - | 5 | 931 | |||
| BC108351.1 | - | 5 | 959 | 6312193 | ||

Pitpnc1
Genomic Map