siRNA Details
| Gene Symbol: Pitpnc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145823.2 | NP_665822.1 | 6 | 1219 | ||
| GenBank mRNA | AB077281.1 | - | 6 | 1200 | ||
| AB077282.1 | - | 6 | 497 | |||
| AK003844.1 | - | 5 | 467 | |||
| AK021206.1 | - | 6 | 432 | |||
| AK153625.1 | - | 6 | 1220 | |||
| AK171340.1 | - | 6 | 1246 | |||
| AK171918.1 | - | 6 | 1216 | |||
| BC082333.1 | - | 6 | 1051 | |||
| BC108351.1 | - | 6 | 1079 | 6312193 | ||

Pitpnc1
Genomic Map