siRNA Details
| Gene Symbol: Rnf180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027934.2 | NP_082210.1 | 4 | 889 | ||
| GenBank mRNA | AK013941.1 | - | 4 | 755 | ||
| AK032259.1 | - | 4 | 858 | |||
| AK136632.1 | - | 4 | 890 | |||
| AK151379.1 | - | 4 | 864 | |||
| AK152404.1 | - | 4 | 864 | |||
| AK157911.1 | - | 829 | ||||
| BC046775.1 | - | 4 | 910 | 5708557 | ||
| BC075700.1 | - | 4 | 910 | 5708557 | ||

Rnf180
Genomic Map