siRNA Details
| Gene Symbol: 1700006A11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027939.1 | NP_082215.1 | 8 | 1196 | ||
| GenBank mRNA | AK005660.1 | - | 7 | 981 | ||
| AK132965.1 | - | 8 | 1208 | |||
| BC137977.1 | - | 8 | 1196 | 8860996 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700006A11Rik| Gene Symbol: 1700006A11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027939.1 | NP_082215.1 | 8 | 1196 | ||
| GenBank mRNA | AK005660.1 | - | 7 | 981 | ||
| AK132965.1 | - | 8 | 1208 | |||
| BC137977.1 | - | 8 | 1196 | 8860996 | ||