siRNA Details
| Gene Symbol: 1700007B14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164235.1 | NP_001157707.1 | 12 | 1601 | ||
| NM_027944.1 | NP_082220.1 | 11 | 1483 | |||
| GenBank mRNA | AK005693.1 | - | 11 | 1484 | ||
| AK077102.1 | - | 13 | 1340 | |||
| BC117767.1 | - | 12 | 1504 | 40090978 | ||
| BC117768.1 | - | 11 | 1386 | 40090984 | ||

1700007B14Rik
Genomic Map